(3,5-Difluorphenyl)methansulfonsäurefluorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3,5-Difluoro-α-toluenesulphonyl chloride
PC50011-1G
PC50011-250MG
APOSPC50011-1GEA
87
EUR
APOSPC50011-1G
APOSPC50011-250MG
(3,5-Difluorphenyl)methansulfonsäurefluorid 97%
(3,5-Difluorphenyl)methansulfonsäurefluorid
Formel:
C₇H₅ClF₂O₂S Molecular Weight: 226.63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09743349 CAS-Nummer: 163295-74-7 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...