2-Nitro-4-(trifluoromethoxy)phenyl trifluoromethanesulphonate

Lieferant: Apollo Scientific
Gefahr

PC500116-1G PC500116-250MG
APOSPC500116-1GEA 0 EUR
APOSPC500116-1G APOSPC500116-250MG
2-Nitro-4-(trifluoromethoxy)phenyl trifluoromethanesulphonate
2-Nitro-4-(trifluoromethoxy)phenyl trifluoromethanesulphonate
Formel: C₈H₃F₆NO₆S
Lagertemperatur: Raumtemperatur
MDL: MFCD25541876
CAS-Nummer: 955395-91-2
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR