3-Fluor-2-methoxyphenol

Lieferant: Apollo Scientific
Achtung

PC500162-1G
APOSPC500162-1GEA 286 EUR
APOSPC500162-1G
3-Fluor-2-methoxyphenol
3-Fluor-2-methoxyphenol
Formel: C₇H₇FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12756463
CAS-Nummer: 96994-70-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR