2-Fluoro-3-(trifluoromethoxy)phenol

Lieferant: Apollo Scientific
Achtung

PC500165-1G
APOSPC500165-1GEA 364 EUR
APOSPC500165-1G
2-Fluoro-3-(trifluoromethoxy)phenol
2-Fluoro-3-(trifluoromethoxy)phenol
Formel: C₇H₄F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22208493
CAS-Nummer: 1334164-87-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR