4-Fluor-3-(trifluormethoxy)phenol

Lieferant: Apollo Scientific
Achtung

PC500166-1G PC500166-5G PC500166-25G
APOSPC500166-1GEA 0 EUR
APOSPC500166-1G APOSPC500166-5G APOSPC500166-25G
4-Fluor-3-(trifluormethoxy)phenol
4-Fluor-3-(trifluormethoxy)phenol
Formel: C₇H₄F₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06660257
CAS-Nummer: 886501-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR