4-Brom-2-(trifluormethoxy)phenol 97+%
Lieferant: Apollo Scientific
|
Achtung
|
PC500169-10G
PC500169-1G
PC500169-5G
APOSPC500169-10GEA
481
EUR
APOSPC500169-10G
APOSPC500169-1G
APOSPC500169-5G
4-Brom-2-(trifluormethoxy)phenol 97+%
4-Brom-2-(trifluormethoxy)phenol
Formel:
C₇H₄BrF₃O₂ Schmelzpunkt: 51…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09037798 CAS-Nummer: 690264-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...