2-Chloro-6-(trifluoromethyl)phenol

Lieferant: Apollo Scientific

Synonyme: 2-Trifluoromethyl-6-chlorophenol

PC500170-1G
APOSPC500170-1GEA 358 EUR
APOSPC500170-1G
2-Chloro-6-(trifluoromethyl)phenol
2-Chloro-6-(trifluoromethyl)phenol
Formel: C₇H₄ClF₃O
Molecular Weight: 196,554 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00673986
CAS-Nummer: 106877-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR