3-Chloro-4-(trifluoromethyl)phenol 98%

Lieferant: Apollo Scientific
Achtung

PC500172-1G PC500172-250MG PC500172-5G
APOSPC500172-1GEA 0 EUR
APOSPC500172-1G APOSPC500172-250MG APOSPC500172-5G
3-Chloro-4-(trifluoromethyl)phenol 98%
3-Chloro-4-(trifluoromethyl)phenol
Formel: C₇H₄ClF₃O
Lagertemperatur: Kühlschrank
MDL: MFCD11849933
CAS-Nummer: 37900-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR