4-Chloro-2-(trifluoromethyl)phenol

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-trifluoromethylphenol

PC500173-1G
APOSPC500173-1GEA 87 EUR
APOSPC500173-1G
4-Chloro-2-(trifluoromethyl)phenol
4-Chloro-2-(trifluoromethyl)phenol
Formel: C₇H₄ClF₃O
Molecular Weight: 196,554 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060673
CAS-Nummer: 53903-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR