2-Methyl-3-(trifluormethyl)phenylacetonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC500182-1G
PC500182-5G
APOSPC500182-1GEA
0
EUR
APOSPC500182-1G
APOSPC500182-5G
2-Methyl-3-(trifluormethyl)phenylacetonitril 98%
2-Methyl-3-(trifluormethyl)phenylacetonitril
Formel:
C₁₀H₈F₃N Schmelzpunkt: 51…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832278 CAS-Nummer: 1000515-00-3 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...