1-Brom-2,3-difluor-4-nitrobenzol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Difluoro-4-bromonitrobenzene

PC500190-1G PC500190-5G
APOSPC500190-1GEA 60.9 EUR
APOSPC500190-1G APOSPC500190-5G
1-Brom-2,3-difluor-4-nitrobenzol
1-Bromo-2,3-difluoro-4-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09839215
CAS-Nummer: 1003708-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR