6-Bromo-7-fluoro-1H-indazole

Lieferant: Apollo Scientific

PC500199-1G
APOSPC500199-1GEA 102 EUR
APOSPC500199-1G
6-Bromo-7-fluoro-1H-indazole
6-Bromo-7-fluoro-1H-indazole
Formel: C₇H₄BrFN₂
Molecular Weight: 215,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23704110
CAS-Nummer: 1427396-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR