2,4-Difluoro-5-hydroxybenzotrifluoride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,4-Difluoro-5-(trifluoromethyl)phenol
PC500224-1G
PC500224-250MG
APOSPC500224-1GEA
551
EUR
APOSPC500224-1G
APOSPC500224-250MG
2,4-Difluoro-5-hydroxybenzotrifluoride
2,4-Difluoro-5-hydroxybenzotrifluoride
Formel:
C₇H₃F₅O Lagertemperatur: Raumtemperatur |
MDL:
MFCD27937185 CAS-Nummer: 1632283-20-5 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...