2,4-Difluoro-5-(trifluoromethyl)benzoic acid 95%

Lieferant: Apollo Scientific
Achtung

PC500226-1G PC500226-250MG PC500226-5G
APOSPC500226-1GEA 0 EUR
APOSPC500226-1G APOSPC500226-250MG APOSPC500226-5G
2,4-Difluoro-5-(trifluoromethyl)benzoic acid 95%
2,4-Difluoro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD01631617
CAS-Nummer: 261945-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR