2,6-Difluoro-3-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC500239-1G PC500239-5G PC500239-250MG
APOSPC500239-1GEA 0 EUR
APOSPC500239-1G APOSPC500239-5G APOSPC500239-250MG
2,6-Difluoro-3-(trifluoromethyl)benzoic acid
2,6-Difluoro-3-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03412249
CAS-Nummer: 1048921-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR