4-Bromo-2-fluoro-5-nitrophenol 95%

Lieferant: Apollo Scientific
Achtung

PC500262-1G
APOSPC500262-1GEA 82.8 EUR
APOSPC500262-1G
4-Bromo-2-fluoro-5-nitrophenol 95%
4-Bromo-2-fluoro-5-nitrophenol
Formel: C₆H₃BrFNO₃
Molecular Weight: 235.995 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100973
CAS-Nummer: 661463-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR