2,6-Difluoro-4-(trifluoromethyl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

PC500301-1G PC500301-250MG PC500301-5G
APOSPC500301-1GEA 0 EUR
APOSPC500301-1G APOSPC500301-250MG APOSPC500301-5G
2,6-Difluoro-4-(trifluoromethyl)benzaldehyde
2,6-Difluoro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₃F₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD12404959
CAS-Nummer: 134099-34-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR