2,6-Difluoro-4-(trifluoromethyl)benzoic acid

Lieferant: Apollo Scientific
Achtung

PC500305-1G PC500305-250MG
APOSPC500305-1GEA 0 EUR
APOSPC500305-1G APOSPC500305-250MG
2,6-Difluoro-4-(trifluoromethyl)benzoic acid
2,6-Difluoro-4-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27975469
CAS-Nummer: 1309597-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR