4-Fluoro-5-iodo-2-methylanisole
Lieferant: Apollo Scientific
|
Achtung
|
PC500306-1G
PC500306-250MG
PC500306-5G
APOSPC500306-1GEA
288
EUR
APOSPC500306-1G
APOSPC500306-250MG
APOSPC500306-5G
4-Fluoro-5-iodo-2-methylanisole
4-Fluoro-5-iodo-2-methylanisole
Formel:
C₈H₈FIO Molecular Weight: 266.051 g/mol Schmelzpunkt: 40…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19440267 CAS-Nummer: 741716-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...