4-Fluoro-5-iodo-2-methylanisole

Lieferant: Apollo Scientific
Achtung

PC500306-1G PC500306-250MG PC500306-5G
APOSPC500306-1GEA 288 EUR
APOSPC500306-1G APOSPC500306-250MG APOSPC500306-5G
4-Fluoro-5-iodo-2-methylanisole
4-Fluoro-5-iodo-2-methylanisole
Formel: C₈H₈FIO
Molecular Weight: 266.051 g/mol
Schmelzpunkt: 40…42 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19440267
CAS-Nummer: 741716-19-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR