3-Fluor-5-iodbenzotrifluorid
Lieferant: Apollo Scientific
Synonyme:
1-Fluoro-3-iodo-5-(trifluoromethyl)benzene
PC500391-100MG
PC500391-1G
PC500391-250MG
APOSPC500391-100MGEA
178
EUR
APOSPC500391-100MG
APOSPC500391-1G
APOSPC500391-250MG
3-Fluor-5-iodbenzotrifluorid
3-Fluor-5-iodbenzotrifluorid
Formel:
C₇H₃F₄I Molecular Weight: 290 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09800718 CAS-Nummer: 1027513-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...