4-Bromo-2-iodo-1-(trifluoromethyl)benzene

Lieferant: Apollo Scientific

Synonyme: 4-Bromo-2-iodobenzotrifluoride

PC500392-5G
APOSPC500392-5GEA 77 EUR
APOSPC500392-5G
4-Bromo-2-iodo-1-(trifluoromethyl)benzene
4-Bromo-2-iodo-1-(trifluoromethyl)benzene
Formel: C₇H₃BrF₃I
Lagertemperatur: Raumtemperatur
MDL: MFCD21603817
CAS-Nummer: 1256945-00-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR