Methyl 2-chloro-3-fluorobenzoate

Lieferant: Apollo Scientific

PC500399-1G PC500399-5G PC500399-25G
APOSPC500399-1GEA 179 EUR
APOSPC500399-1G APOSPC500399-5G APOSPC500399-25G
Methyl 2-chloro-3-fluorobenzoate
Methyl 2-chloro-3-fluorobenzoate
Formel: C₈H₆ClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10566821
CAS-Nummer: 647020-70-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR