3-Fluoro-2-(trifluoromethyl)phenylacetylene 95+%

Lieferant: Apollo Scientific

PC500411-1G
APOSPC500411-1GEA 873 EUR
APOSPC500411-1G
3-Fluoro-2-(trifluoromethyl)phenylacetylene 95+%
3-Fluoro-2-(trifluoromethyl)phenylacetylene
Formel: C₉H₄F₄
Lagertemperatur: Raumtemperatur
MDL: MFCD27995605
CAS-Nummer: 1823923-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR