4-Bromo-5-chloro-2-fluorobenzoic acid 98%

Lieferant: Apollo Scientific
Achtung

PC500421-1G PC500421-250MG PC500421-5G
APOSPC500421-1GEA 60.9 EUR
APOSPC500421-1G APOSPC500421-250MG APOSPC500421-5G
4-Bromo-5-chloro-2-fluorobenzoic acid 98%
4-Bromo-5-chloro-2-fluorobenzoic acid
Formel: C₇H₃BrClFO₂
Molecular Weight: 253.453 g/mol
Schmelzpunkt: 152…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19686184
CAS-Nummer: 1349708-91-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR