4-Bromo-5-chloro-2-fluorobenzoic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC500421-1G
PC500421-250MG
PC500421-5G
APOSPC500421-1GEA
60.9
EUR
APOSPC500421-1G
APOSPC500421-250MG
APOSPC500421-5G
4-Bromo-5-chloro-2-fluorobenzoic acid 98%
4-Bromo-5-chloro-2-fluorobenzoic acid
Formel:
C₇H₃BrClFO₂ Molecular Weight: 253.453 g/mol Schmelzpunkt: 152…154 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19686184 CAS-Nummer: 1349708-91-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...