5-Bromo-2-fluoro-4-methoxybenzaldehyde

Lieferant: Apollo Scientific

PC500445-1G
APOSPC500445-1GEA 314 EUR
APOSPC500445-1G
5-Bromo-2-fluoro-4-methoxybenzaldehyde
5-Bromo-2-fluoro-4-methoxybenzaldehyde
Formel: C₈H₆BrFO₂
Molecular Weight: 233,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11847135
CAS-Nummer: 473417-48-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR