2-Fluoro-4-hydroxyphenylacetic acid
Lieferant: Apollo Scientific
PC500449-1G
APOSPC500449-1GEA
109
EUR
APOSPC500449-1G
2-Fluoro-4-hydroxyphenylacetic acid
2-Fluoro-4-hydroxyphenylacetic acid
Formel:
C₈H₇FO₃ Molecular Weight: 170,1400 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027748 CAS-Nummer: 68886-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...