5-Bromo-2-fluoro-4-methoxybenzoic acid

Lieferant: Apollo Scientific
Achtung

PC500451-1G
APOSPC500451-1GEA 277 EUR
APOSPC500451-1G
5-Bromo-2-fluoro-4-methoxybenzoic acid
5-Bromo-2-fluoro-4-methoxybenzoic acid
Formel: C₈H₆BrFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD20486270
CAS-Nummer: 949014-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR