3-(Bromomethyl)-5-fluorobenzoic acid

Lieferant: Apollo Scientific
Gefahr

PC500474-1G PC500474-250MG PC500474-5G
APOSPC500474-1GEA 319 EUR
APOSPC500474-1G APOSPC500474-250MG APOSPC500474-5G
3-(Bromomethyl)-5-fluorobenzoic acid
3-(Bromomethyl)-5-fluorobenzoic acid
Formel: C₈H₆BrFO₂
Lagertemperatur: Kühlschrank
MDL: MFCD18378634
CAS-Nummer: 1379351-68-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR