5-(Bromomethyl)-2-fluorobenzoic acid

Lieferant: Apollo Scientific
Gefahr

PC500479-1G PC500479-250MG PC500479-5G
APOSPC500479-1GEA 328 EUR
APOSPC500479-1G APOSPC500479-250MG APOSPC500479-5G
5-(Bromomethyl)-2-fluorobenzoic acid
5-(Bromomethyl)-2-fluorobenzoic acid
Formel: C₈H₆BrFO₂
Lagertemperatur: Kühlschrank
MDL: MFCD06208849
CAS-Nummer: 773100-76-8
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR