3-Bromo-6-chloro-2-fluoroaniline 98%

Lieferant: Apollo Scientific
Achtung

PC500489-1G PC500489-250MG
APOSPC500489-1GEA 0 EUR
APOSPC500489-1G APOSPC500489-250MG
3-Bromo-6-chloro-2-fluoroaniline 98%
3-Bromo-6-chloro-2-fluoroaniline
Formel: C₆H₄BrClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD27934056
CAS-Nummer: 943830-81-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR