2'-Amino-2,2,2-trifluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Trifluoroacetyl)aniline, 1-(2-Aminophenyl)-2,2,2-trifluoroethan-1-one

PC50050-100MG
APOSPC50050-100MGEA 203 EUR
APOSPC50050-100MG
2'-Amino-2,2,2-trifluoroacetophenone
2'-Amino-2,2,2-trifluoroacetophenone
Formel: C₈H₆F₃NO
Lagertemperatur: Kühlschrank
MDL: MFCD09863840
CAS-Nummer: 351002-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR