3,5-Dichlor-4-fluorbenzaldehyd

Lieferant: Apollo Scientific

PC500504-1G
APOSPC500504-1GEA 94.4 EUR
APOSPC500504-1G
3,5-Dichlor-4-fluorbenzaldehyd
3,5-Dichloro-4-fluorobenzaldehyde
Formel: C₇H₃Cl₂FO
Lagertemperatur: Raumtemperatur
MDL: MFCD11845864
CAS-Nummer: 117820-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR