3-(Brommethyl)-4-fluorbenzonitril

Lieferant: Apollo Scientific
Gefahr

PC500514-1G PC500514-250MG PC500514-5G
APOSPC500514-1GEA 241 EUR
APOSPC500514-1G APOSPC500514-250MG APOSPC500514-5G
3-(Brommethyl)-4-fluorbenzonitril
3-(Brommethyl)-4-fluorbenzonitril
Formel: C₈H₅BrFN
Lagertemperatur: Kühlschrank
CAS-Nummer: 856935-35-8
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR