2'-Amino-5'-chloro-2,2,2-trifluoroacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
PC50052-1G
APOSPC50052-1GEA
201
EUR
APOSPC50052-1G
2'-Amino-5'-chloro-2,2,2-trifluoroacetophenone
2'-Amino-5'-chloro-2,2,2-trifluoroacetophenone
Formel:
C₈H₅ClF₃NO Lagertemperatur: Kühlschrank |
MDL:
MFCD08458216 CAS-Nummer: 154598-53-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...