4-Bromo-2-(trifluoromethyl)phenylacetonitrile
Lieferant: Apollo Scientific
PC500531-1G
APOSPC500531-1GEA
0
EUR
APOSPC500531-1G
4-Bromo-2-(trifluoromethyl)phenylacetonitrile
4-Bromo-2-(trifluoromethyl)phenylacetonitrile
Formel:
C₉H₅BrF₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185431 CAS-Nummer: 877131-92-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...