2'-Amino-3'-methoxy-2,2,2-trifluoroacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxy-6-(trifluoroacetyl)aniline, 2-Amino-3-(trifluoroacetyl)anisole, 1-(2-Amino-3-methoxyphenyl)-2,2,2-trifluoroethan-1-one

PC50054-1G PC50054-250MG
APOSPC50054-1GEA 0 EUR
APOSPC50054-1G APOSPC50054-250MG
2'-Amino-3'-methoxy-2,2,2-trifluoroacetophenone
2'-Amino-3'-methoxy-2,2,2-trifluoroacetophenone
Formel: C₉H₈F₃NO₂
Molecular Weight: 219.161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13173364
CAS-Nummer: 489429-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR