Methyl 4-bromo-2-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific
Achtung

PC500545-1G PC500545-5G PC500545-25G
APOSPC500545-1GEA 0 EUR
APOSPC500545-1G APOSPC500545-5G APOSPC500545-25G
Methyl 4-bromo-2-(trifluoromethyl)benzoate
Methyl 4-bromo-2-(trifluoromethyl)benzoate
Formel: C₉H₆BrF₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12761599
CAS-Nummer: 957207-58-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR