2-(Bromomethyl)-6-fluorobenzonitrile 97%

Lieferant: Apollo Scientific
Gefahr

PC500549-1G
APOSPC500549-1GEA 85.7 EUR
APOSPC500549-1G
2-(Bromomethyl)-6-fluorobenzonitrile 97%
2-(Bromomethyl)-6-fluorobenzonitrile
Formel: C₈H₅BrFN
Lagertemperatur: Raumtemperatur
MDL: MFCD18394042
CAS-Nummer: 1261686-95-6
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR