4-(Bromomethyl)-3-fluorobenzaldehyde

Lieferant: Apollo Scientific
Gefahr

PC500558-1G PC500558-250MG PC500558-5G PC500558-25G
APOSPC500558-1GEA 0 EUR
APOSPC500558-1G APOSPC500558-250MG APOSPC500558-5G APOSPC500558-25G
4-(Bromomethyl)-3-fluorobenzaldehyde
4-(Bromomethyl)-3-fluorobenzaldehyde
Formel: C₈H₆BrFO
Lagertemperatur: Kühlschrank
MDL: MFCD28096625
CAS-Nummer: 1416980-24-9
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR