4-Fluoro-5-(trifluoromethoxy)benzene-1,2-diamine 95%

Lieferant: Apollo Scientific
Achtung

PC500567-1G PC500567-250MG
APOSPC500567-1GEA 0 EUR
APOSPC500567-1G APOSPC500567-250MG
4-Fluoro-5-(trifluoromethoxy)benzene-1,2-diamine 95%
4-Fluoro-5-(trifluoromethoxy)benzene-1,2-diamine
Formel: C₇H₆F₄N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD28096583
CAS-Nummer: 1804386-40-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR