Potassium (2,2-dimethylprop-1-yl)trifluoroborate 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC50057-1G
APOSPC50057-1GEA
66.7
EUR
APOSPC50057-1G
Potassium (2,2-dimethylprop-1-yl)trifluoroborate 98%
Potassium (2,2-dimethylprop-1-yl)trifluoroborate
Formel:
C₅H₁₁BF₃·K Lagertemperatur: Raumtemperatur |
MDL:
MFCD09271780 CAS-Nummer: 1150655-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...