3-(Bromomethyl)-2-fluorobenzaldehyde

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Fluoro-3-formylbenzyl bromide

PC500570-1G PC500570-250MG
APOSPC500570-1GEA 543 EUR
APOSPC500570-1G APOSPC500570-250MG
3-(Bromomethyl)-2-fluorobenzaldehyde
3-(Bromomethyl)-2-fluorobenzaldehyde
Formel: C₈H₆BrFO
Molecular Weight: 217.035 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18378584
CAS-Nummer: 1379307-88-6
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR