5-(Bromomethyl)-2-fluorobenzaldehyde

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Fluoro-3-formylbenzyl bromide, α-Bromo-6-fluoro-m-tolualdehyde

PC500573-1G PC500573-250MG PC500573-5G
APOSPC500573-1GEA 435 EUR
APOSPC500573-1G APOSPC500573-250MG APOSPC500573-5G
5-(Bromomethyl)-2-fluorobenzaldehyde
5-(Bromomethyl)-2-fluorobenzaldehyde
Formel: C₈H₆BrFO
Molecular Weight: 217.035 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28096626
CAS-Nummer: 1820717-85-8
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR