Kalium-1,3-dimethyluracil-5-trifluoroborat

Lieferant: Apollo Scientific

PC50058-1G
APOSPC50058-1GEA 292 EUR
APOSPC50058-1G
Kalium-1,3-dimethyluracil-5-trifluoroborat
Kalium-1,3-dimethyluracil-5-trifluoroborat
Formel: C₆H₇BF₃KN₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD09271782
CAS-Nummer: 1150654-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR