4-(Bromomethyl)-3-(trifluoromethyl)benzoic acid, tech

Lieferant: Apollo Scientific
Achtung

PC500593-1G PC500593-250MG PC500593-5G
APOSPC500593-1GEA 0 EUR
APOSPC500593-1G APOSPC500593-250MG APOSPC500593-5G
4-(Bromomethyl)-3-(trifluoromethyl)benzoic acid, tech
4-(Bromomethyl)-3-(trifluoromethyl)benzoic acid
Formel: C₉H₆BrF₃O₂
Schmelzpunkt: 140…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD22492213
CAS-Nummer: 859213-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR