Potassium [(benzyloxy)methyl]trifluoroborate

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium ((benzyloxy)methyl)trifluoroborate

PC50064-5G
APOSPC50064-5GEA 133 EUR
APOSPC50064-5G
Potassium [(benzyloxy)methyl]trifluoroborate
Potassium [(benzyloxy)methyl]trifluoroborate
Formel: C₈H₉BF₃O·K
Molecular Weight: 228,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052815
CAS-Nummer: 1027642-25-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR