4-Bromo-2,3-difluorobenzylamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-Bromo-2,3-difluorophenyl)methanamine hydrochloride

PC500666-250MG
APOSPC500666-250MGEA 106 EUR
APOSPC500666-250MG
4-Bromo-2,3-difluorobenzylamine hydrochloride 97%
4-Bromo-2,3-difluorobenzylamine hydrochloride
Formel: C₇H₆BrF₂N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD12546123
CAS-Nummer: 1980086-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR