4'-Iodo-3'-(trifluoromethyl)acetophenone 95%

Lieferant: Apollo Scientific
Achtung

PC500668-1G PC500668-250MG PC500668-5G
APOSPC500668-1GEA 0 EUR
APOSPC500668-1G APOSPC500668-250MG APOSPC500668-5G
4'-Iodo-3'-(trifluoromethyl)acetophenone 95%
4'-Iodo-3'-(trifluoromethyl)acetophenone
Formel: C₉H₆F₃IO
Lagertemperatur: Raumtemperatur
MDL: MFCD28103433
CAS-Nummer: 1823318-98-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR